Assay Details
Gene Symbol: P2rx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080780.2 | NP_542958.2 | 1 - 2 | 317 | 80 | |
XM_006246566.2 | XP_006246628.1 | 1 - 2 | 397 | 80 | ||
GenBank mRNA | X92069.1 | - | 1 - 2 | 317 | 80 | |
X97328.1 | - | 1 - 2 | 183 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
