Assay Details
Gene Symbol: Daxx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080891.2 | NP_543167.2 | 7 - 8 | 2229 | 80 | |
XM_006256109.1 | XP_006256171.1 | 6 - 7 | 3104 | 80 | ||
XM_006256110.2 | XP_006256172.1 | 7 - 8 | 2230 | 80 | ||
GenBank mRNA | AB064671.1 | - | 6 - 7 | 2136 | 80 | |
BC128729.1 | - | 7 - 8 | 2229 | 80 | ||
FQ223571.1 | - | 7 - 8 | 2200 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
