Assay Details
Gene Symbol: Arf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257320.2 | XP_006257382.1 | 1 - 2 | 194 | 100 | |
GenBank mRNA | L12382.1 | - | 1 - 2 | 98 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the L12382.1, XM_006257320.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
