Assay Details
Gene Symbol: Dynlrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131910.3 | NP_571985.1 | 3 - 4 | 365 | 102 | |
GenBank mRNA | AF073839.2 | - | 3 - 4 | 288 | 102 | |
AY026512.1 | - | 3 - 4 | 365 | 102 | ||
BC058437.1 | - | 3 - 4 | 306 | 6921076 | 102 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
