Assay Details
Gene Symbol: Necab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133415.1 | NP_596906.1 | 1 - 2 | 163 | 120 | |
GenBank mRNA | AF193757.1 | - | 1 - 2 | 163 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P269425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160234 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P269425 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P269425 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P269425 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P269425 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702161812 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66607 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56746 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702161812 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10808319 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808313 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808320 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808316 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808318 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808323 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808317 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808314 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808322 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808321 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808315 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808324 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10808312 | - | - |
Rat Genome 230 2.0 Array | 1368443_at | - | - |
Rat Genome 230 2.0 Array | 1368443_PM_at | - | - |
Rat Genome 230 2.0 Array | 1396751_at | - | - |
Rat Genome 230 2.0 Array | 1396751_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1396751_at | - | - |
Affymetrix HT Rat Focus Array | 1368443_at | - | - |
Affymetrix HT Rat Focus Array | 1368443_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1396751_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1900005862.rn.1 | TC1900000709.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1900005863.rn.1 | TC1900000709.rn.1 | RTA TC |
Rat Expression Set 230 | 1368443_PM_at | - | - |
Rat Expression Set 230 | 1368443_at | - | - |
Rat Expression Set 230 | 1396751_at | - | - |
Rat Expression Set 230 | 1396751_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808312 | - | - |
Rat Genome U34 Set | rc_AI044096_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22413535 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364683 | - | - |
RatRef-12 Expression BeadChip | ILMN_55705 | - | - |
