Assay Details
Gene Symbol: Cabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133529.2 | NP_598213.1 | 1 - 2 | 47 | 66 | |
GenBank mRNA | AJ315657.1 | - | 1 - 2 | 47 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
negative regulation of cell communication by electrical coupling
negative regulation of protein import into nucleus
regulation of synaptic plasticity
regulation of voltage-gated calcium channel activity
negative regulation of voltage-gated calcium channel activity
calcium ion binding
protein binding
nuclear localization sequence binding
protein domain specific binding
type 3 metabotropic glutamate receptor binding
protein homodimerization activity
ion channel binding
calcium-dependent protein binding
multicellular organism development
negative regulation of cell communication by electrical coupling
negative regulation of protein import into nucleus
regulation of synaptic plasticity
regulation of voltage-gated calcium channel activity
negative regulation of voltage-gated calcium channel activity
calcium ion binding
protein binding
nuclear localization sequence binding
protein domain specific binding
type 3 metabotropic glutamate receptor binding
protein homodimerization activity
ion channel binding
calcium-dependent protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P142922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62816 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P142922 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P142922 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P142922 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P524820 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 60681 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35718 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50657 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46012 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10759017 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759014 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759011 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759015 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759013 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759016 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759012 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759010 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10759009 | - | - |
Rat Genome 230 2.0 Array | 1396749_at | - | - |
Rat Genome 230 2.0 Array | 1369886_a_at | - | - |
Rat Genome 230 2.0 Array | 1396749_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369886_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1369886_a_at | - | - |
Affymetrix HT Rat Focus Array | 1396749_at | - | - |
Affymetrix HT Rat Focus Array | 1396749_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369886_PM_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1200006883.rn.1 | TC1200000782.rn.1 | RTA TC |
Rat Expression Set 230 | 1369886_a_at | - | - |
Rat Expression Set 230 | 1369886_PM_a_at | - | - |
Rat Expression Set 230 | 1396749_at | - | - |
Rat Expression Set 230 | 1396749_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759009 | - | - |
Rat Genome U34 Set | Y17048_g_at | - | - |
Rat Genome U34 Set | Y17048_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21666104 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360836 | - | - |
RatRef-12 Expression BeadChip | ILMN_48596 | - | - |
