Assay Details
Gene Symbol: Slc31a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133600.1 | NP_598284.1 | 1 - 2 | 112 | 97 | |
GenBank mRNA | AF268030.1 | - | 1 - 2 | 112 | 97 | |
BC078745.1 | - | 1 - 2 | 74 | 97 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF268030.1, BC078745.1, NM_133600.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
