Assay Details
Gene Symbol: Elovl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134382.1 | NP_599209.1 | 5 - 6 | 622 | 80 | |
XM_006243417.1 | XP_006243479.1 | 6 - 7 | 787 | 80 | ||
GenBank mRNA | AB071985.1 | - | 5 - 6 | 622 | 80 | |
HQ404314.1 | - | 5 - 6 | 622 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
