Assay Details
Gene Symbol: Epcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138541.1 | NP_612550.1 | 1 - 2 | 142 | 120 | |
GenBank mRNA | AJ001044.1 | - | 1 - 2 | 197 | 120 | |
BC061787.1 | - | 1 - 2 | 146 | 120 | ||
BC072691.1 | - | 1 - 2 | 166 | 7098920 | 120 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AJ001044.1, BC072691.1, NM_138541.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
