Assay Details
Gene Symbol: Rabggtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138708.2 | NP_619715.1 | 5 - 6 | 590 | 120 | |
GenBank mRNA | BC060536.1 | - | 614 | 120 | ||
BC078683.1 | - | 5 - 6 | 579 | 120 | ||
FQ216744.1 | - | 594 | 120 | |||
FQ224712.1 | - | 593 | 120 | |||
FQ229109.1 | - | 594 | 120 | |||
L10416.1 | - | 5 - 6 | 590 | 120 | ||
S62097.1 | - | 5 - 6 | 590 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
