Assay Details
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138831.1 | NP_620186.1 | 4 - 5 | 1278 | 69 | |
XM_008772974.1 | XP_008771196.1 | 4 - 5 | 1317 | 69 | ||
GenBank mRNA | AB047324.1 | - | 4 - 5 | 1278 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P226315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926700 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64500 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P226315 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P226315 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P226315 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P226315 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86089 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 54373 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8180 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27286 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64666 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 752 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96503 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42060 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8472 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7713 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 93920 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10833641 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833642 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833636 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833638 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833640 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833637 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10833635 | - | - |
Rat Genome 230 2.0 Array | 1370548_at | - | - |
Rat Genome 230 2.0 Array | 1370548_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370548_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370548_at | - | - |
Affymetrix Rat Clariom S Assay | PSR2000013673.rn.1 | TC2000001686.rn.1 | RTA TC |
Rat Expression Set 230 | 1370548_PM_at | - | - |
Rat Expression Set 230 | 1370548_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833635 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21949794 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365262 | - | - |
RatRef-12 Expression BeadChip | ILMN_53436 | - | - |
