Assay Details
Gene Symbol: Syt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138835.1 | NP_620190.1 | 1 - 2 | 199 | 94 | |
GenBank mRNA | U71294.1 | - | 1 - 2 | 199 | 94 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_138835.1, U71294.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P089356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169700 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16876 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P089356 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P089356 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P089356 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P144188 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700521229 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701746526 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47296 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12444 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701746526 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700521229 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10727592 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10727590 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10727595 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10727591 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10727589 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10727594 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10727593 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10727596 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10727588 | - | - |
Rat Genome 230 2.0 Array | 1388060_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388060_at | - | - |
Affymetrix HT Rat Focus Array | 1388060_at | - | - |
Affymetrix HT Rat Focus Array | 1388060_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100056332.rn.1 | TC0100007074.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100056338.rn.1 | TC0100007074.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100056331.rn.1 | TC0100007074.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100056333.rn.1 | TC0100007074.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100056335.rn.1 | TC0100007074.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100056337.rn.1 | TC0100007074.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100056334.rn.1 | TC0100007074.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100056336.rn.1 | TC0100007074.rn.1 | RTA TC |
Rat Expression Set 230 | 1388060_PM_at | - | - |
Rat Expression Set 230 | 1388060_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10727588 | - | - |
Rat Genome U34 Set | U71294_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22157012 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354055 | - | - |
RatRef-12 Expression BeadChip | ILMN_68586 | - | - |
