Assay Details
Gene Symbol: Haus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138864.2 | NP_620219.2 | 2 - 3 | 219 | 128 | |
GenBank mRNA | AF092207.1 | - | 215 | 128 | ||
BC127484.1 | - | 2 - 3 | 219 | 128 |
Gene Symbol: NEWGENE_619909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772211.1 | XP_008770433.1 | 244 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
