Assay Details
Gene Symbol: Bmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139258.1 | NP_640351.1 | 4 - 5 | 703 | 62 | |
XM_006234723.2 | XP_006234785.1 | 4 - 5 | 741 | 62 | ||
GenBank mRNA | AF506761.1 | - | 4 - 5 | 703 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
