Assay Details
Gene Symbol: Faim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144756.2 | NP_653357.1 | 1 - 2 | 94 | 74 | |
GenBank mRNA | AF044201.1 | - | 1 - 2 | 43 | 74 | |
BC087606.1 | - | 1 - 2 | 94 | 7314068 | 74 |
Gene Symbol: LOC102551901 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242293.2 | XP_006242355.1 | 1 - 2 | 320 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF044201.1, BC087606.1, NM_144756.2, XM_006242293.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
