Assay Details
Gene Symbol: Filip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145682.1 | NP_663715.1 | 2 - 3 | 344 | 90 | |
GenBank mRNA | AB055759.1 | - | 2 - 3 | 344 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P596942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16144 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P596942 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P596942 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P596942 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1000722 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P596942 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2147 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12822 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95938 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10918957 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918958 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918956 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918959 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10918955 | - | - |
Rat Genome 230 2.0 Array | 1379463_at | - | - |
Rat Genome 230 2.0 Array | 1375089_at | - | - |
Rat Genome 230 2.0 Array | 1379463_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375089_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375089_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375089_at | - | - |
Affymetrix HT Rat Focus Array | 1379463_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379463_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800024096.rn.1 | TC0800003006.rn.1 | RTA TC |
Rat Expression Set 230 | 1375089_PM_at | - | - |
Rat Expression Set 230 | 1375089_at | - | - |
Rat Expression Set 230 | 1379463_PM_at | - | - |
Rat Expression Set 230 | 1379463_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918955 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21849570 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376850 | - | - |
