Assay Details
Gene Symbol: Yif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172017.2 | NP_742014.2 | 1 - 2 | 183 | 56 | |
GenBank mRNA | BC158578.1 | - | 1 - 2 | 164 | 7387164 | 56 |
BF521694.1 | - | 1 - 2 | 190 | 56 | ||
CV074812.1 | - | 1 - 2 | 163 | 56 | ||
FQ214813.1 | - | 1 - 2 | 286 | 56 | ||
FQ219390.1 | - | 1 - 2 | 178 | 56 | ||
FQ220202.1 | - | 1 - 2 | 180 | 56 | ||
U96490.1 | - | 1 - 2 | 124 | 56 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC158578.1, NM_172017.2, U96490.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
