Assay Details
Gene Symbol: Stox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF548355.1 | - | 1 | 927 | 93 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P022790 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P022785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108683 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P022785 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P022790 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P022790 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P022785 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P022785 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P022790 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P505964 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P133226 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10788171 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788174 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788177 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788173 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788176 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788172 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10788175 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10788170 | - | - |
Rat Genome 230 2.0 Array | 1376815_at | - | - |
Rat Genome 230 2.0 Array | 1378798_PM_at | - | - |
Rat Genome 230 2.0 Array | 1384808_PM_at | - | - |
Rat Genome 230 2.0 Array | 1376815_PM_at | - | - |
Rat Genome 230 2.0 Array | 1384808_at | - | - |
Rat Genome 230 2.0 Array | 1378798_at | - | - |
Affymetrix HT Rat Focus Array | 1384808_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1378798_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376815_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376815_at | - | - |
Affymetrix HT Rat Focus Array | 1384808_at | - | - |
Affymetrix HT Rat Focus Array | 1378798_at | - | - |
Rat Expression Set 230 | 1376815_at | - | - |
Rat Expression Set 230 | 1376815_PM_at | - | - |
Rat Expression Set 230 | 1378798_PM_at | - | - |
Rat Expression Set 230 | 1384808_PM_at | - | - |
Rat Expression Set 230 | 1384808_at | - | - |
Rat Expression Set 230 | 1378798_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788170 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788175 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21238048 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374371 | - | - |
