Assay Details
Gene Symbol: Dpy30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173117.2 | NP_775140.2 | 1 - 2 | 106 | 96 | |
GenBank mRNA | AY129401.1 | - | 1 - 2 | 106 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY129401.1, NM_173117.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
