Assay Details
Gene Symbol: Tph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173839.2 | NP_776211.1 | 8 - 9 | 1171 | 64 | |
XM_006241352.2 | XP_006241414.1 | 8 - 9 | 1227 | 64 | ||
XM_008765357.1 | XP_008763579.1 | 8 - 9 | 1233 | 64 | ||
GenBank mRNA | AY098915.1 | - | 8 - 9 | 1171 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
circadian rhythm
aromatic amino acid family metabolic process
response to activity
response to nutrient levels
serotonin biosynthetic process
response to estrogen
response to glucocorticoid
response to calcium ion
oxidation-reduction process
cellular response to lithium ion
molecular_function
tryptophan 5-monooxygenase activity
iron ion binding
amino acid binding
circadian rhythm
aromatic amino acid family metabolic process
response to activity
response to nutrient levels
serotonin biosynthetic process
response to estrogen
response to glucocorticoid
response to calcium ion
oxidation-reduction process
cellular response to lithium ion
molecular_function
tryptophan 5-monooxygenase activity
iron ion binding
amino acid binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P311178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53294 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P311178 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P311178 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P311178 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P311178 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64692 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33201 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84543 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98617 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78860 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18712 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52658 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58520 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70041 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10902372 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902366 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902369 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902364 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902371 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902373 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902370 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902368 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902367 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902374 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902365 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10902363 | - | - |
Affymetrix Rat Clariom S Assay | PSR0700021058.rn.1 | TC0700002714.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021061.rn.1 | TC0700002714.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021057.rn.1 | TC0700002714.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021063.rn.1 | TC0700002714.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021064.rn.1 | TC0700002714.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021059.rn.1 | TC0700002714.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021062.rn.1 | TC0700002714.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021067.rn.1 | TC0700002714.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021069.rn.1 | TC0700002714.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021060.rn.1 | TC0700002714.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021068.rn.1 | TC0700002714.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10902363 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21578095 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357302 | - | - |
RatRef-12 Expression BeadChip | ILMN_65392 | - | - |
