Assay Details
Gene Symbol: Cd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175577.3 | NP_783167.2 | 3 - 4 | 543 | 96 | |
XM_006231021.2 | XP_006231083.1 | 3 - 4 | 650 | 96 | ||
XM_008760218.1 | XP_008758440.1 | 3 - 4 | 650 | 96 | ||
XM_008760219.1 | XP_008758441.1 | 3 - 4 | 649 | 96 | ||
GenBank mRNA | AF488727.1 | - | 3 - 4 | 482 | 96 | |
AY683561.1 | - | 3 - 4 | 465 | 96 | ||
BC089767.1 | - | 3 - 4 | 543 | 96 |
Gene Symbol: LOC100911366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760255.1 | XP_008758477.1 | 2 - 3 | 580 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
