Assay Details
Gene Symbol: Ppil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175707.3 | NP_783638.1 | 4 - 5 | 485 | 63 | |
XM_006244964.2 | XP_006245026.1 | 4 - 5 | 555 | 63 | ||
GenBank mRNA | AF315802.1 | - | 4 - 5 | 273 | 63 | |
BC087645.1 | - | 4 - 5 | 485 | 63 | ||
FQ224392.1 | - | 4 - 5 | 282 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
