Assay Details
Gene Symbol: Cryl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175757.2 | NP_786933.1 | 7 - 8 | 907 | 75 | |
XM_006252058.2 | XP_006252120.1 | 5 - 6 | 1428 | 75 | ||
GenBank mRNA | AY040223.1 | - | 7 - 8 | 907 | 75 | |
BC078685.1 | - | 7 - 8 | 881 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P065190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60065 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P065190 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P065190 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P065190 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1002360 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41628 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104524 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97377 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88958 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42872 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91194 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102260 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4562 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10784128 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784133 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784129 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784134 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784132 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784127 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784131 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784126 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784130 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10784125 | - | - |
Rat Genome 230 2.0 Array | 1376051_at | - | - |
Rat Genome 230 2.0 Array | 1376051_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376051_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376051_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1500011768.rn.1 | TC1500001776.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500011767.rn.1 | TC1500001776.rn.1 | RTA TC |
Rat Expression Set 230 | 1376051_PM_at | - | - |
Rat Expression Set 230 | 1376051_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784125 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22103877 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365959 | - | - |
RatRef-12 Expression BeadChip | ILMN_51307 | - | - |
