Assay Details
Gene Symbol: Slc1a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175758.3 | NP_786934.1 | 2 - 3 | 1006 | 73 | |
GenBank mRNA | AJ132846.1 | - | 2 - 3 | 737 | 73 | |
BC080242.1 | - | 2 - 3 | 1006 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
