Assay Details
Gene Symbol: G6pc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176077.3 | NP_788266.2 | 4 - 5 | 708 | 101 | |
GenBank mRNA | AY186240.1 | - | 5 - 6 | 538 | 101 | |
BC078689.1 | - | 4 - 5 | 708 | 101 | ||
FQ231610.1 | - | 4 - 5 | 604 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
