Assay Details
Gene Symbol: Arhgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107747.2 | NP_001101217.2 | 2 - 3 | 264 | 71 | |
XM_006234551.1 | XP_006234613.1 | 3 - 4 | 301 | 71 | ||
XM_006234552.2 | XP_006234614.1 | 3 - 4 | 331 | 71 | ||
XM_006234553.2 | XP_006234615.1 | 3 - 4 | 301 | 71 | ||
XM_006234554.1 | XP_006234616.1 | 4 - 5 | 343 | 71 | ||
XM_006234556.1 | XP_006234618.1 | 3 - 4 | 278 | 71 | ||
XM_006234557.2 | XP_006234619.1 | 3 - 4 | 308 | 71 | ||
XM_006234558.2 | XP_006234620.1 | 2 - 3 | 249 | 71 | ||
XM_006234560.2 | XP_006234622.1 | 2 - 3 | 290 | 71 | ||
GenBank mRNA | CK480577.1 | - | 2 - 3 | 264 | 71 | |
FQ224002.1 | - | 2 - 3 | 224 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
