Assay Details
Gene Symbol: Hnrnpa2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104613.1 | NP_001098083.1 | 2 - 3 | 150 | 108 | |
GenBank mRNA | AB006815.1 | - | 1 - 2 | 114 | 108 | |
AB006816.1 | - | 2 - 3 | 150 | 108 | ||
AB006817.1 | - | 1 - 2 | 114 | 108 | ||
AB006818.1 | - | 2 - 3 | 150 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
