Assay Details
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012522.2 | NP_036654.2 | 4 - 5 | 586 | 77 | |
GenBank mRNA | D01098.1 | - | 4 - 5 | 587 | 77 | |
M88344.1 | - | 4 - 5 | 735 | 77 | ||
M88345.1 | - | 1 - 2 | 999 | 77 | ||
M88346.1 | - | 3 - 4 | 481 | 77 | ||
M88347.1 | - | 3 - 4 | 481 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
