Assay Details
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012522.2 | NP_036654.2 | 7 - 8 | 867 | 60 | |
GenBank mRNA | D01098.1 | - | 7 - 8 | 868 | 60 | |
FQ211441.1 | - | 7 - 8 | 926 | 60 | ||
M88344.1 | - | 7 - 8 | 1016 | 60 | ||
M88345.1 | - | 4 - 5 | 1280 | 60 | ||
M88346.1 | - | 6 - 7 | 762 | 60 | ||
M88347.1 | - | 6 - 7 | 762 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
