Assay Details
Gene Symbol: Odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239906.2 | 10 - 11 | 1389 | 96 | ||
XM_006239907.2 | XP_006239969.1 | 10 - 11 | 1379 | 96 | ||
XM_006239908.2 | XP_006239970.1 | 9 - 10 | 1295 | 96 | ||
XM_008764573.1 | 10 - 11 | 1362 | 96 | |||
GenBank mRNA | BC078882.1 | - | 8 - 9 | 1067 | 7115603 | 96 |
J04791.1 | - | 10 - 11 | 1183 | 96 | ||
M16982.1 | - | 10 - 11 | 1447 | 96 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
