Assay Details
Gene Symbol: Pklr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012624.3 | NP_036756.3 | 2 - 3 | 208 | 60 | |
XM_006232593.2 | XP_006232655.1 | 2 - 3 | 339 | 60 | ||
GenBank mRNA | BC161827.1 | - | 1 - 2 | 198 | 60 | |
M11709.1 | - | 2 - 3 | 209 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
