Assay Details
Gene Symbol: Camk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012727.3 | NP_036859.2 | 10 - 11 | 923 | 90 | |
XM_006254523.2 | XP_006254585.1 | 9 - 10 | 865 | 90 | ||
GenBank mRNA | BC128706.1 | - | 10 - 11 | 923 | 90 | |
M63334.1 | - | 817 | 90 | |||
M64757.1 | - | 8 - 9 | 913 | 90 | ||
S65840.1 | - | 9 - 10 | 848 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
