Assay Details
Gene Symbol: Ucp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013167.2 | NP_037299.1 | 6 - 7 | 937 | 86 | |
GenBank mRNA | AB006614.1 | - | 5 - 6 | 809 | 86 | |
AF030163.1 | - | 5 - 6 | 824 | 86 | ||
AF035943.1 | - | 5 - 6 | 820 | 86 | ||
BC072546.1 | - | 6 - 7 | 937 | 7105743 | 86 | |
U92069.1 | - | 6 - 7 | 944 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
