Assay Details
Gene Symbol: Aif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017196.3 | NP_058892.1 | 3 - 4 | 250 | 67 | |
XM_006256065.2 | XP_006256127.1 | 5 - 6 | 400 | 67 | ||
GenBank mRNA | AB000818.1 | - | 3 - 4 | 248 | 67 | |
D82069.1 | - | 5 - 6 | 405 | 67 | ||
FM053942.1 | - | 3 - 4 | 237 | 67 | ||
FQ231499.1 | - | 3 - 4 | 248 | 67 | ||
U17919.1 | - | 3 - 4 | 222 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
