Assay Details
Gene Symbol: Slc7a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017353.1 | NP_059049.1 | 4 - 5 | 893 | 71 | |
GenBank mRNA | AB015432.1 | - | 4 - 5 | 893 | 71 | |
U00995.1 | - | 2 - 3 | 218 | 71 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the U00995.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
