Assay Details
Gene Symbol: Milr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021585.1 | NP_067596.1 | 6 - 7 | 774 | 126 | |
XM_008768389.1 | XP_008766611.1 | 4 - 5 | 715 | 126 | ||
GenBank mRNA | U39546.1 | - | 6 - 7 | 774 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
