Assay Details
Gene Symbol: Fgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239078.2 | XP_006239140.1 | 10 - 11 | 1389 | 69 | |
XM_006239079.2 | XP_006239141.1 | 11 - 12 | 1421 | 69 | ||
XM_006239080.2 | XP_006239142.1 | 11 - 12 | 1421 | 69 | ||
XM_006239081.2 | XP_006239143.1 | 11 - 12 | 1502 | 69 | ||
XM_006239082.2 | XP_006239144.1 | 10 - 11 | 1433 | 69 | ||
GenBank mRNA | X57018.1 | - | 11 - 12 | 1430 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
