Assay Details
Gene Symbol: Gk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024381.2 | NP_077357.2 | 5 - 6 | 510 | 132 | |
XM_006256987.2 | XP_006257049.1 | 5 - 6 | 544 | 132 | ||
XM_006256988.2 | XP_006257050.1 | 5 - 6 | 544 | 132 | ||
XM_008773274.1 | XP_008771496.1 | 6 - 7 | 609 | 132 | ||
XM_008773275.1 | XP_008771497.1 | 6 - 7 | 609 | 132 | ||
XM_008773276.1 | XP_008771498.1 | 6 - 7 | 609 | 132 | ||
XM_008773278.1 | XP_008771500.1 | 6 - 7 | 608 | 132 | ||
GenBank mRNA | D16102.1 | - | 5 - 6 | 513 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
