Assay Details
Gene Symbol: Gk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024381.2 | NP_077357.2 | 7 - 8 | 758 | 114 | |
XM_006256987.2 | XP_006257049.1 | 7 - 8 | 792 | 114 | ||
XM_006256988.2 | XP_006257050.1 | 7 - 8 | 792 | 114 | ||
XM_008773274.1 | XP_008771496.1 | 8 - 9 | 857 | 114 | ||
XM_008773275.1 | XP_008771497.1 | 8 - 9 | 857 | 114 | ||
XM_008773276.1 | XP_008771498.1 | 8 - 9 | 857 | 114 | ||
XM_008773278.1 | XP_008771500.1 | 8 - 9 | 856 | 114 | ||
GenBank mRNA | D16102.1 | - | 7 - 8 | 761 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
