Assay Details
Gene Symbol: Hmgcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024386.1 | NP_077362.1 | 4 - 5 | 361 | 70 | |
GenBank mRNA | BC061797.1 | - | 4 - 5 | 363 | 70 | |
FQ209392.1 | - | 4 - 5 | 359 | 70 | ||
FQ209616.1 | - | 4 - 5 | 365 | 70 | ||
FQ234382.1 | - | 4 - 5 | 359 | 70 | ||
Y10054.1 | - | 4 - 5 | 361 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
