Assay Details
Gene Symbol: Hmgcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024386.1 | NP_077362.1 | 5 - 6 | 515 | 93 | |
GenBank mRNA | BC061797.1 | - | 5 - 6 | 517 | 5598033 | 93 |
FQ209392.1 | - | 5 - 6 | 513 | 93 | ||
FQ209616.1 | - | 5 - 6 | 519 | 93 | ||
FQ234382.1 | - | 5 - 6 | 513 | 93 | ||
Y10054.1 | - | 5 - 6 | 515 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
