Assay Details
Gene Symbol: Hpgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024390.2 | NP_077366.2 | 6 - 7 | 677 | 104 | |
GenBank mRNA | BC062399.1 | - | 6 - 7 | 677 | 104 | |
U44750.1 | - | 6 - 7 | 693 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
prostaglandin metabolic process
transforming growth factor beta receptor signaling pathway
female pregnancy
parturition
ovulation
negative regulation of cell cycle
thrombin-activated receptor signaling pathway
ductus arteriosus closure
catalytic activity
prostaglandin E receptor activity
15-hydroxyprostaglandin dehydrogenase (NAD+) activity
NAD binding
NAD+ binding
prostaglandin metabolic process
transforming growth factor beta receptor signaling pathway
female pregnancy
parturition
ovulation
negative regulation of cell cycle
thrombin-activated receptor signaling pathway
ductus arteriosus closure
catalytic activity
prostaglandin E receptor activity
15-hydroxyprostaglandin dehydrogenase (NAD+) activity
NAD binding
NAD+ binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P11278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7373 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11278 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11278 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P11278 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11278 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72422 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1500 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 67964 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10791529 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791527 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791526 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791525 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791528 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791530 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791523 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791524 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10791522 | - | - |
Rat Genome 230 2.0 Array | 1376191_at | - | - |
Rat Genome 230 2.0 Array | 1387631_PM_at | - | - |
Rat Genome 230 2.0 Array | 1387631_at | - | - |
Rat Genome 230 2.0 Array | 1376191_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376191_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376191_at | - | - |
Affymetrix HT Rat Focus Array | 1387631_at | - | - |
Affymetrix HT Rat Focus Array | 1387631_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1600011879.rn.1 | TC1600001566.rn.1 | RTA TC |
Rat Expression Set 230 | 1387631_PM_at | - | - |
Rat Expression Set 230 | 1376191_at | - | - |
Rat Expression Set 230 | 1387631_at | - | - |
Rat Expression Set 230 | 1376191_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791522 | - | - |
Rat Genome U34 Set | U44750_at | - | - |
Rat Genome U34 Set | rc_AA848820_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21233763 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367170 | - | - |
RatRef-12 Expression BeadChip | ILMN_53529 | - | - |
