Assay Details
Gene Symbol: Gnai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031035.3 | NP_112297.1 | 1 - 2 | 269 | 58 | |
XM_006243858.1 | XP_006243920.1 | 1 - 2 | 299 | 58 | ||
GenBank mRNA | AY899210.1 | - | 1 - 2 | 117 | 58 | |
AY899211.1 | - | 1 - 2 | 120 | 58 | ||
BC078806.1 | - | 1 - 2 | 221 | 58 | ||
CO394955.1 | - | 1 - 2 | 269 | 58 | ||
DQ120471.1 | - | 1 - 2 | 178 | 58 | ||
DQ120472.1 | - | 1 - 2 | 178 | 58 | ||
M12672.1 | - | 1 - 2 | 165 | 58 | ||
M17528.1 | - | 1 - 2 | 267 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
