Assay Details
Gene Symbol: Gnai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031035.3 | NP_112297.1 | 6 - 7 | 876 | 94 | |
XM_006243858.1 | XP_006243920.1 | 6 - 7 | 906 | 94 | ||
GenBank mRNA | AY899210.1 | - | 6 - 7 | 724 | 94 | |
BC078806.1 | - | 6 - 7 | 828 | 94 | ||
DQ120471.1 | - | 6 - 7 | 785 | 94 | ||
DQ120472.1 | - | 6 - 7 | 785 | 94 | ||
M12672.1 | - | 6 - 7 | 772 | 94 | ||
M17528.1 | - | 6 - 7 | 874 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
