Assay Details
Gene Symbol: Gtf2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031041.1 | NP_112303.1 | 4 - 5 | 412 | 84 | |
GenBank mRNA | BC085345.1 | - | 4 - 5 | 417 | 7303747 | 84 |
FQ229769.1 | - | 4 - 5 | 400 | 84 | ||
FQ231337.1 | - | 4 - 5 | 442 | 84 | ||
X65948.1 | - | 4 - 5 | 412 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
