Assay Details
Gene Symbol: Gtf2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031041.1 | NP_112303.1 | 5 - 6 | 542 | 130 | |
GenBank mRNA | BC085345.1 | - | 5 - 6 | 547 | 130 | |
FQ229769.1 | - | 5 - 6 | 530 | 130 | ||
X65948.1 | - | 5 - 6 | 542 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
