Assay Details
Gene Symbol: Rnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031097.1 | NP_112359.1 | 9 - 10 | 1655 | 131 | |
GenBank mRNA | AY724503.1 | - | 9 - 10 | 1830 | 131 | |
BC061718.1 | - | 9 - 10 | 1688 | 131 | ||
D87515.1 | - | 9 - 10 | 1652 | 131 | ||
U61696.1 | - | 9 - 10 | 1655 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
