Assay Details
Gene Symbol: Ssr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031120.2 | NP_112382.2 | 279 | 88 | ||
GenBank mRNA | AY724496.1 | - | 287 | 88 | ||
BC062015.1 | - | 279 | 88 | |||
Z14030.1 | - | 287 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
