Assay Details
Gene Symbol: Tgfbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031132.3 | NP_112394.3 | 4 - 5 | 1507 | 104 | |
XM_008766690.1 | XP_008764912.1 | 5 - 6 | 2063 | 104 | ||
GenBank mRNA | GU067683.1 | - | 4 - 5 | 1256 | 104 | |
L09653.1 | - | 4 - 5 | 1314 | 104 | ||
S67770.1 | - | 4 - 5 | 1507 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
