Assay Details
Gene Symbol: Slc20a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031148.1 | NP_112410.1 | 1 - 2 | 400 | 85 | |
XM_006234991.2 | XP_006235053.1 | 1 - 2 | 624 | 85 | ||
XM_006234992.2 | XP_006235054.1 | 1 - 2 | 624 | 85 | ||
GenBank mRNA | AB000489.1 | - | 1 - 2 | 440 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
